Assay Details
Target Gene Details
Entrez Gene ID: | 55502 |
Gene Name: | hes family bHLH transcription factor 6 |
Gene Aliases: |
C-HAIRY1, HES-6, bHLHb41, bHLHc23 |
Location: |
Chr.2:238238267-238240124 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HES6 | NM_001142853.2 | 4 | 568 | NP_001136325.1 |
| NM_001282434.1 | 4 | 534 | NP_001269363.1 | |
| NM_018645.5 | 4 | 574 | NP_061115.2 | |
| AB035179.1 | 4 | 355 | BAA96082.1 | |
| AF260237.1 | 4 | 415 | AAK51634.1 | |
| AK075040.1 | 3 | 723 | BAC11368.1 | |
| AK293111.1 | 4 | 488 | ||
| BC007939.2 | 4 | 479 | AAH07939.1 | |
| BE796220.1 | 4 | 574 | ||
| DB495731.1 | 4 | 443 | ||
| DT217800.1 | 4 | 491 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584761 | Chr.2:238212011 - 238246412 on Build GRCh38 | Gain |
|
| nsv584764 | Chr.2:238238530 - 238240682 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
| dgv7349n54 | Chr.2:238238228 - 238241307 on Build GRCh38 | Loss |
|
| esv21505 | Chr.2:238238959 - 238241354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map