Assay Details
Target Gene Details
Entrez Gene ID: | 4849 |
Gene Name: | CCR4-NOT transcription complex subunit 3 |
Gene Aliases: |
LENG2, NOT3, NOT3H |
Location: |
Chr.19:54137689-54155708 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT3 | NM_014516.3 | NP_055331.1 | ||
| XM_005278279.1 | XP_005278336.1 | |||
| XM_005278280.2 | XP_005278337.1 | |||
| XM_005278281.1 | XP_005278338.1 | |||
| XM_005278282.2 | XP_005278339.1 | |||
| XM_011526992.1 | XP_011525294.1 | |||
| AB014591.1 | ||||
| AF180474.1 | AAF29828.1 | |||
| AF211967.1 | 2 | 177 | ||
| AI660119.1 | ||||
| AK160386.1 | 16 | 2631 | BAD18729.1 | |
| AK300434.1 | ||||
| AL133647.1 | CAB63766.1 | |||
| BC016474.1 | AAH16474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644782 | Chr.19:54138064 - 54154026 on Build GRCh38 | Gain |
|
| esv3644783 | Chr.19:54151022 - 54159664 on Build GRCh38 | Gain |
|
| nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss |
|
| nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map