Assay Details
Target Gene Details
Entrez Gene ID: | 100128568 |
Gene Name: | uncharacterized LOC100128568 |
Gene Aliases: |
- |
Location: |
Chr.19:5978403-6020363 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128568 | NR_046376.1 |
Target Gene Details
Entrez Gene ID: | 5990 |
Gene Name: | regulatory factor X2 |
Gene Aliases: |
- |
Location: |
Chr.19:5993164-6110653 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX2 | NM_000635.3 | 15 | 2045 | NP_000626.2 |
| NM_134433.2 | 14 | 1970 | NP_602309.1 | |
| XM_011528171.2 | 16 | 2011 | XP_011526473.1 | |
| XM_011528172.1 | 15 | 1868 | XP_011526474.1 | |
| XM_017027107.1 | 14 | 1895 | XP_016882596.1 | |
| XM_017027108.1 | 13 | 1820 | XP_016882597.1 | |
| AK024288.1 | 12 | 1584 | ||
| AK093977.1 | 7 | 2600 | ||
| AK291196.1 | 14 | 1849 | ||
| AK302661.1 | 9 | 1033 | ||
| AK303321.1 | 12 | 1354 | ||
| BC028579.1 | 15 | 1957 | AAH28579.1 | |
| BC071571.1 | 15 | 1926 | AAH71571.1 | |
| BX648130.1 | 15 | 1880 | ||
| X76091.1 | 15 | 1935 | CAA53705.1 |
More Information
Set Membership: |
|

Genomic Map