Assay Details
Target Gene Details
Entrez Gene ID: | 83758 |
Gene Name: | retinol binding protein 5 |
Gene Aliases: |
CRBP-III, CRBP3, CRBPIII, HRBPiso |
Location: |
Chr.12:7119106-7130272 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP5 | XM_017020002.1 | XP_016875491.1 | ||
| AF212239.1 | 1 | 122 | AAK14925.1 | |
| AK096947.1 | 1 | 137 | ||
| AK223378.1 | 1 | 137 | BAD97098.1 | |
| AK311977.1 | 1 | 135 | ||
| AY007436.1 | 1 | 118 | AAG09617.1 | |
| BC029355.1 | 1 | 139 | AAH29355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818882 | Chr.12:7128721 - 7149386 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map