Assay Details
Target Gene Details
Entrez Gene ID: | 152503 |
Gene Name: | SH3 domain containing 19 |
Gene Aliases: |
EBP, EVE1, Kryn, SH3P19 |
Location: |
Chr.4:151120281-151228030 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595690 | Chr.4:150867803 - 151188400 on Build GRCh38 | Gain |
|
| esv3602674 | Chr.4:151094056 - 151193747 on Build GRCh38 | Gain |
|
| dgv5436n100 | Chr.4:150743286 - 151150724 on Build GRCh38 | Gain |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
| nsv526976 | Chr.4:150754667 - 151133921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map