Assay Details
Target Gene Details
Entrez Gene ID: | 23089 |
Gene Name: | paternally expressed 10 |
Gene Aliases: |
EDR, HB-1, MEF3L, Mar2, Mart2, RGAG3 |
Location: |
Chr.7:94656325-94669695 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEG10 | NM_001040152.1 | 2 | 2718 | NP_001035242.1 |
| NM_001172437.2 | 2 | 2729 | NP_001165908.1 | |
| NM_001172438.2 | 2 | 2729 | NP_001165909.1 | |
| NM_001184961.1 | 2 | 2718 | NP_001171890.1 | |
| NM_001184962.1 | 2 | 2718 | NP_001171891.1 | |
| NM_015068.3 | 2 | 2718 | NP_055883.2 | |
| AB028974.1 | 1 | 2291 | BAA83003.2 | |
| AB049150.1 | 2 | 2497 | BAB68387.1 | |
| AB049834.1 | 1 | 2356 | BAB43951.1 | |
| AF216076.1 | 1 | 2249 | ||
| AK025529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422248 | Chr.7:94543833 - 94955318 on Build GRCh38 | Duplication |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map