Assay Details
Target Gene Details
Entrez Gene ID: | 4239 |
Gene Name: | microfibrillar associated protein 4 |
Gene Aliases: |
- |
Location: |
Chr.17:19383442-19387219 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFAP4 | NM_001198695.1 | 6 | 1362 | NP_001185624.1 |
| NM_002404.2 | 6 | 1253 | NP_002395.1 | |
| AK293056.1 | 6 | 1224 | ||
| AK304528.1 | 6 | 1333 | ||
| BC062415.1 | 5 | 1190 | AAH62415.1 | |
| L38486.1 | 5 | 1193 | AAB00968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
| nsv953845 | Chr.17:19377788 - 19387787 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map