Assay Details
Target Gene Details
Entrez Gene ID: | 102191832 |
Gene Name: | dual endothelin-1(VEGFsp)/angiotensin II receptor pseudogene |
Gene Aliases: |
DEspR |
Location: |
Chr.4:152337655-152338096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEAR | NR_104273.1 | |||
| EF212178.1 |
Target Gene Details
Entrez Gene ID: | 55294 |
Gene Name: | F-box and WD repeat domain containing 7 |
Gene Aliases: |
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4 |
Location: |
Chr.4:152321258-152536095 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4553 | Chr.4:152323453 - 152349980 on Build GRCh38 | Insertion |
|
| dgv5442n100 | Chr.4:152331622 - 152388386 on Build GRCh38 | Loss |
|
| nsv516218 | Chr.4:152308716 - 152388386 on Build GRCh38 | Loss |
|
| dgv5440n100 | Chr.4:152321467 - 152392321 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map