Assay Details
Target Gene Details
Entrez Gene ID: | 283748 |
Gene Name: | phospholipase A2 group IVD |
Gene Aliases: |
cPLA2delta |
Location: |
Chr.15:42067562-42094559 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4D | NM_178034.3 | 17 | 1869 | NP_828848.3 |
| XM_011521467.2 | 16 | 1700 | XP_011519769.1 | |
| AB090876.1 | 17 | 1872 | BAC67158.1 | |
| BC034571.1 | 3 | 306 | AAH34571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv1043641 | Chr.15:42044386 - 42089042 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map