Assay Details
Target Gene Details
Entrez Gene ID: | 84085 |
Gene Name: | F-box protein 30 |
Gene Aliases: |
Fbx30 |
Location: |
Chr.6:145798135-145864409 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO30 | NM_032145.4 | 3 | 3448 | NP_115521.3 |
| XM_005267159.3 | 3 | 3287 | XP_005267216.1 | |
| XM_011536177.2 | 3 | 8307 | XP_011534479.1 | |
| XM_017011353.1 | 3 | 3287 | XP_016866842.1 | |
| AF248640.1 | 2 | 2329 | AAK30299.1 | |
| AK055919.1 | 1 | 1172 | ||
| AK123026.1 | 2 | 1889 | ||
| AK124138.1 | 2 | 2648 | ||
| BC024326.1 | 3 | 3416 | AAH24326.3 |
Target Gene Details
Entrez Gene ID: | 100507557 |
Gene Name: | uncharacterized LOC100507557 |
Gene Aliases: |
- |
Location: |
Chr.6:145734869-145886585 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507557 | NR_038246.1 | |||
| AK124757.1 | ||||
| DB465589.1 |
More Information
Set Membership: |
|

Genomic Map