Assay Details
Target Gene Details
Entrez Gene ID: | 55255 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
MSTP048 |
Location: |
Chr.5:77426652-77620611 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR41 | NM_018268.3 | 13 | 2344 | NP_060738.2 |
| XM_005248551.4 | 11 | 2137 | XP_005248608.1 | |
| XM_011543505.2 | 13 | 2106 | XP_011541807.1 | |
| XM_017009606.1 | 13 | 2754 | XP_016865095.1 | |
| AF115511.1 | 13 | 2092 | AAO06948.1 | |
| AK001766.1 | 13 | 2290 | BAA91895.1 | |
| AK023977.1 | 13 | 2046 | BAB14749.1 | |
| AL834138.1 | 7 | 3025 | CAD38853.2 | |
| BC040241.1 | 13 | 2096 | AAH40241.1 | |
| BP276774.1 | 1 | 200 |

Genomic Map