Assay Details
Target Gene Details
Entrez Gene ID: | 8483 |
Gene Name: | cartilage intermediate layer protein |
Gene Aliases: |
CILP-1, HsT18872 |
Location: |
Chr.15:65195999-65211608 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CILP | NM_003613.3 | NP_003604.3 | ||
| AY358904.1 | AAQ89263.1 | |||
| CA503413.1 |
More Information
Set Membership: |
|

Genomic Map