Assay Details
Target Gene Details
Entrez Gene ID: | 25766 |
Gene Name: | pre-mRNA processing factor 40 homolog B |
Gene Aliases: |
HYPC |
Location: |
Chr.12:49622717-49644669 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map