Assay Details
Target Gene Details
Entrez Gene ID: | 136541 |
Gene Name: | protease, serine 58 |
Gene Aliases: |
PRSS1, TRY1, TRYX3, UNQ2540 |
Location: |
Chr.7:142252138-142258058 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS58 | NM_001001317.4 | NP_001001317.1 | ||
| AK122940.1 | ||||
| AY358487.1 | AAQ88851.1 | |||
| BC035384.1 | AAH35384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035111 | Chr.7:142136794 - 142310364 on Build GRCh38 | Gain |
|
| nsv465131 | Chr.7:142072828 - 142629318 on Build GRCh38 | Loss |
|
| nsv1018286 | Chr.7:142236996 - 142767091 on Build GRCh38 | Gain |
|
| nsv608591 | Chr.7:142185403 - 142323575 on Build GRCh38 | Gain |
|
| esv33962 | Chr.7:142061773 - 142348667 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map