Assay Details
Target Gene Details
Entrez Gene ID: | 221391 |
Gene Name: | opsin 5 |
Gene Aliases: |
GPR136, GRP136, PGR12, TMEM13 |
Location: |
Chr.6:47782037-47831088 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPN5 | NM_181744.3 | 4 | 648 | NP_859528.1 |
| NR_033806.1 | 4 | 665 | ||
| XM_017010409.1 | 2 | 1519 | XP_016865898.1 | |
| XM_017010410.1 | XP_016865899.1 | |||
| XM_017010411.1 | 3 | 756 | XP_016865900.1 | |
| XM_017010412.1 | 3 | 812 | XP_016865901.1 | |
| XM_017010413.1 | 3 | 469 | XP_016865902.1 | |
| XM_017010414.1 | XP_016865903.1 | |||
| XM_017010415.1 | 4 | 665 | XP_016865904.1 | |
| XM_017010416.1 | 4 | 650 | XP_016865905.1 | |
| AK097448.1 | 1 | 155 | ||
| AY255615.1 | 2 | 227 | AAO85127.1 | |
| AY288419.1 | 4 | 611 | AAP72128.1 | |
| AY377391.1 | 4 | 625 | AAR21109.1 | |
| BC126194.1 | 4 | 648 | ||
| BC126198.1 | 4 | 648 | ||
| BX647224.1 | 4 | 607 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map