Assay Details
Target Gene Details
Entrez Gene ID: | 220002 |
Gene Name: | cytochrome b561 family member A3 |
Gene Aliases: |
CYBASC3, LCYTB |
Location: |
Chr.11:61348748-61362299 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB561A3 | NM_001161452.1 | NP_001154924.1 | ||
| NM_001161454.1 | NP_001154926.1 | |||
| NM_001300763.1 | NP_001287692.1 | |||
| NM_153611.4 | NP_705839.3 | |||
| XM_011544821.1 | XP_011543123.1 | |||
| XM_011544823.1 | XP_011543125.1 | |||
| XM_011544824.1 | XP_011543126.1 | |||
| XM_017017345.1 | XP_016872834.1 | |||
| XM_017017346.1 | XP_016872835.1 | |||
| XM_017017347.1 | XP_016872836.1 | |||
| AK056751.1 | ||||
| AK075559.1 | BAC11698.1 | |||
| AK225754.1 | ||||
| AK297063.1 | ||||
| AK297084.1 | ||||
| AK310167.1 | ||||
| AK310976.1 | ||||
| BC004391.1 | AAH04391.1 | |||
| BC014045.2 | AAH14045.2 | |||
| BC047710.1 | AAH47710.1 | |||
| BP366144.1 | ||||
| DA948287.1 | ||||
| DC378209.1 | ||||
| DC389589.1 |
More Information
Set Membership: |
|

Genomic Map