Assay Details
Target Gene Details
Entrez Gene ID: | 10360 |
Gene Name: | nucleophosmin/nucleoplasmin 3 |
Gene Aliases: |
PORMIN, TMEM123 |
Location: |
Chr.10:101781325-101783413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPM3 | NM_006993.2 | NP_008924.1 | ||
| AF081280.1 | AAC31609.1 | |||
| AY049737.1 | AAL12172.1 | |||
| BC041067.1 | AAH41067.1 | |||
| BC054868.1 | AAH54868.1 | |||
| BM765409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951003 | Chr.10:101761444 - 101783043 on Build GRCh38 | Deletion |
|
| dgv171n67 | Chr.10:101769728 - 101817385 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map