Assay Details
Target Gene Details
Entrez Gene ID: | 8872 |
Gene Name: | cell division cycle 123 |
Gene Aliases: |
C10orf7, D123 |
Location: |
Chr.10:12195962-12250590 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC123 | NM_006023.2 | NP_006014.2 | ||
| XM_005252638.3 | XP_005252695.1 | |||
| AK225634.1 | ||||
| AK289362.1 | ||||
| AK308790.1 | ||||
| BC001600.1 | AAH01600.1 | |||
| BC009598.1 | AAH09598.1 | |||
| BP367374.1 | ||||
| CR456966.1 | CAG33247.1 | |||
| D14878.1 | BAA03593.1 | |||
| U27112.1 | AAC34738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891757 | Chr.10:12196611 - 12327862 on Build GRCh38 | Gain |
|
| esv3622363 | Chr.10:12200775 - 12828318 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map