Assay Details
Target Gene Details
Entrez Gene ID: | 9667 |
Gene Name: | scaffold attachment factor B2 |
Gene Aliases: |
- |
Location: |
Chr.19:5586993-5622927 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAFB2 | NM_014649.2 | NP_055464.1 | ||
| XM_011528449.2 | XP_011526751.1 | |||
| XM_011528450.2 | XP_011526752.1 | |||
| XM_017027520.1 | XP_016883009.1 | |||
| XM_017027521.1 | XP_016883010.1 | |||
| XM_017027522.1 | XP_016883011.1 | |||
| D50928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map