Assay Details
Target Gene Details
Entrez Gene ID: | 6366 |
Gene Name: | C-C motif chemokine ligand 21 |
Gene Aliases: |
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4 |
Location: |
Chr.9:34709005-34710167 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL21 | NM_002989.3 | 4 | 571 | NP_002980.1 |
| XM_011518004.2 | 4 | 591 | XP_011516306.1 | |
| AB002409.1 | 4 | 528 | BAA21817.1 | |
| AF001979.1 | 4 | 470 | AAB86594.1 | |
| AK311901.1 | ||||
| AY358887.1 | 4 | 475 | AAQ89246.1 | |
| BC027918.1 | 4 | 477 | AAH27918.1 | |
| BI833188.1 | 4 | 553 | ||
| BP331359.1 | ||||
| CR450326.1 | CAG29322.1 | |||
| U88320.1 | 4 | 484 | AAB91454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv509301 | Chr.9:34703177 - 34799269 on Build GRCh38 | Insertion |
|
| nsv614157 | Chr.9:34695472 - 34737831 on Build GRCh38 | Loss |
|
| nsv831544 | Chr.9:34650003 - 34818951 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map