Assay Details
Target Gene Details
Entrez Gene ID: | 8634 |
Gene Name: | RNA 3'-terminal phosphate cyclase |
Gene Aliases: |
RPC, RTC1, RTCD1 |
Location: |
Chr.1:100266158-100292769 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTCA | NM_001130841.1 | 1 | 180 | NP_001124313.1 |
| NM_003729.3 | 1 | 180 | NP_003720.1 | |
| XM_005271298.4 | 1 | 99 | XP_005271355.1 | |
| XM_005271299.4 | 1 | 151 | XP_005271356.1 | |
| AK292366.1 | 1 | 128 | ||
| AK299793.1 | 1 | 180 | ||
| AK307445.1 | 1 | 100 | ||
| AK309394.1 | 1 | 153 | ||
| AU310497.1 | 1 | 180 | ||
| BC012604.1 | 1 | 94 | AAH12604.1 | |
| DA361597.1 | 1 | 122 | ||
| Y11651.1 | 1 | 131 | CAA72364.1 |
More Information
Set Membership: |
|

Genomic Map