Assay Details
Target Gene Details
Entrez Gene ID: | 55020 |
Gene Name: | tetratricopeptide repeat domain 38 |
Gene Aliases: |
LL22NC03-5H6.5 |
Location: |
Chr.22:46267964-46294008 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC38 | NM_017931.2 | NP_060401.2 | ||
| XM_011530259.1 | XP_011528561.1 | |||
| XM_011530260.2 | XP_011528562.1 | |||
| AK000706.1 | BAA91331.1 | |||
| AK300606.1 | ||||
| BC018918.2 | AAH18918.2 | |||
| DA762669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829293 | Chr.22:46241054 - 46741639 on Build GRCh38 | Gain |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
| nsv589136 | Chr.22:46251091 - 46313984 on Build GRCh38 | Gain |
|
| nsv428393 | Chr.22:46259046 - 46302079 on Build GRCh38 | Gain |
|

Genomic Map