Assay Details
Target Gene Details
Entrez Gene ID: | 221927 |
Gene Name: | BRCA1 associated ATM activator 1 |
Gene Aliases: |
BAAT1, C7orf27, RMFSL |
Location: |
Chr.7:2537810-2555758 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRAT1 | NM_152743.3 | NP_689956.2 | ||
| XM_005249643.4 | XP_005249700.1 | |||
| XM_011515177.2 | XP_011513479.1 | |||
| XM_011515178.1 | XP_011513480.1 | |||
| XM_011515179.2 | XP_011513481.1 | |||
| XM_011515181.2 | XP_011513483.1 | |||
| XM_011515182.2 | XP_011513484.1 | |||
| XM_011515183.1 | XP_011513485.1 | |||
| XM_011515184.2 | XP_011513486.1 | |||
| XM_011515186.2 | XP_011513488.1 | |||
| XM_017011833.1 | XP_016867322.1 | |||
| XM_017011834.1 | XP_016867323.1 | |||
| XM_017011835.1 | XP_016867324.1 | |||
| XM_017011836.1 | XP_016867325.1 | |||
| AK024482.1 | BAB15772.1 | |||
| AK131083.1 | 5 | 2404 | BAC85133.1 | |
| AK131097.1 | 1 | 2040 | BAC85147.1 | |
| AK311144.1 | 2 | 452 | ||
| BC007209.1 | AAH07209.1 | |||
| BC015632.2 | AAH15632.2 | |||
| BC023561.2 | AAH23561.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823981 | Chr.7:2507328 - 2543718 on Build GRCh38 | Gain |
|
| nsv830887 | Chr.7:2507249 - 2621396 on Build GRCh38 | Loss |
|
| nsv950792 | Chr.7:2418066 - 2544866 on Build GRCh38 | Deletion |
|
| nsv605862 | Chr.7:2516523 - 2604885 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113638677,rs73287544] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map