Assay Details
Target Gene Details
Entrez Gene ID: | 8631 |
Gene Name: | src kinase associated phosphoprotein 1 |
Gene Aliases: |
HEL-S-81p, SCAP1, SKAP55 |
Location: |
Chr.17:48133438-48430291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKAP1 | NM_001075099.1 | NP_001068567.1 | ||
| NM_003726.3 | NP_003717.3 | |||
| XM_005257755.3 | XP_005257812.1 | |||
| XM_017025257.1 | XP_016880746.1 | |||
| XM_017025258.1 | XP_016880747.1 | |||
| XM_017025259.1 | XP_016880748.1 | |||
| XM_017025260.1 | XP_016880749.1 | |||
| AK290818.1 | ||||
| BC047870.1 | AAH47870.1 | |||
| EU668346.1 | ||||
| Y11215.1 | CAA72101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469808 | Chr.17:47972502 - 48151311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map