Assay Details
Target Gene Details
Entrez Gene ID: | 3567 |
Gene Name: | interleukin 5 |
Gene Aliases: |
EDF, IL-5, TRF |
Location: |
Chr.5:132539194-132556827 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL5 | NM_000879.2 | NP_000870.1 | ||
| XM_005271988.3 | XP_005272045.1 | |||
| XM_011543373.2 | XP_011541675.1 | |||
| XM_011543374.2 | XP_011541676.1 | |||
| BC066282.1 | AAH66282.1 | |||
| DQ655971.2 | ||||
| DQ655974.2 | 1 | 759 | ||
| X04688.1 | CAA28390.1 | |||
| X12705.1 | CAA31210.1 |
More Information
Set Membership: |
|

Genomic Map