Assay Details
Target Gene Details
Entrez Gene ID: | 5338 |
Gene Name: | phospholipase D2 |
Gene Aliases: |
- |
Location: |
Chr.17:4807101-4823432 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD2 | NM_001243108.1 | 20 | 2198 | NP_001230037.1 |
| NM_002663.4 | 20 | 2198 | NP_002654.3 | |
| XM_005256696.2 | 11 | 1258 | XP_005256753.1 | |
| XM_017024764.1 | 20 | 2198 | XP_016880253.1 | |
| AB209374.1 | 15 | 3766 | BAD92611.1 | |
| AF033850.1 | 20 | 2233 | AAD04197.1 | |
| AF035483.1 | 19 | 2072 | AAC24498.1 | |
| AF038440.1 | 20 | 2156 | AAB96655.1 | |
| AF038441.1 | 20 | 2156 | AAB96656.1 | |
| AK314797.1 | 20 | 2116 | ||
| AY178289.1 | 11 | 1163 | AAO21120.1 | |
| BC015033.1 | 20 | 2159 | AAH15033.1 | |
| BC056871.1 | 20 | 2107 | AAH56871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss |
|
| esv3639795 | Chr.17:4812578 - 4820832 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map