Assay Details
Target Gene Details
Entrez Gene ID: | 10531 |
Gene Name: | pitrilysin metallopeptidase 1 |
Gene Aliases: |
MP1, PreP |
Location: |
Chr.10:3137727-3172841 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITRM1 | NM_001242307.1 | NP_001229236.1 | ||
| NM_001242309.1 | NP_001229238.1 | |||
| NM_014889.3 | NP_055704.2 | |||
| XM_005252345.3 | XP_005252402.1 | |||
| XM_017015470.1 | XP_016870959.1 | |||
| XM_017015471.1 | XP_016870960.1 | |||
| XM_017015472.1 | XP_016870961.1 | |||
| XM_017015473.1 | XP_016870962.1 | |||
| AB029027.2 | BAA83056.2 | |||
| AF061243.1 | AAC67244.1 | |||
| AK001183.1 | ||||
| AK002061.1 | ||||
| AK023476.1 | ||||
| AK225084.1 | ||||
| AK294865.1 | ||||
| AK299464.1 | ||||
| AK303406.1 | ||||
| AK315117.1 | ||||
| AL117409.1 | ||||
| BC001150.1 | AAH01150.1 | |||
| BC005025.1 | AAH05025.1 | |||
| BC095422.1 | AAH95422.1 | |||
| BC111987.1 | AAI11988.1 | |||
| BC113369.1 | ||||
| BX649017.1 | ||||
| CR749279.1 | CAH18134.1 |
Target Gene Details
Entrez Gene ID: | 100507034 |
Gene Name: | PITRM1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:3141601-3148629 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITRM1-AS1 | NR_038284.1 | 4 | 1134 | |
| AK097545.1 | 3 | 884 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549814 | Chr.10:3055020 - 3170233 on Build GRCh38 | Loss |
|
| esv3622167 | Chr.10:3137807 - 3185609 on Build GRCh38 | Gain |
|
| dgv932n54 | Chr.10:2382612 - 3253481 on Build GRCh38 | Gain |
|
| esv2730995 | Chr.10:2770220 - 3706473 on Build GRCh38 | Deletion |
|
| nsv428229 | Chr.10:2618610 - 3156581 on Build GRCh38 | Gain |
|
| nsv549818 | Chr.10:3081241 - 3227125 on Build GRCh38 | Gain |
|
| nsv1044106 | Chr.10:3136689 - 3186881 on Build GRCh38 | Loss |
|
| nsv1039231 | Chr.10:3131146 - 3736633 on Build GRCh38 | Loss |
|
| esv2750891 | Chr.10:3098894 - 3164173 on Build GRCh38 | Loss |
|
| nsv549810 | Chr.10:3035272 - 3691194 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map