Assay Details
Target Gene Details
Entrez Gene ID: | 220032 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 4 |
Gene Aliases: |
GDE6 |
Location: |
Chr.11:77216558-77301699 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDPD4 | NM_182833.1 | NP_878253.1 | ||
| XM_011544826.1 | XP_011543128.1 | |||
| XM_011544833.1 | XP_011543135.1 | |||
| XM_011544834.1 | XP_011543136.1 | |||
| XM_017017348.1 | XP_016872837.1 | |||
| XM_017017349.1 | XP_016872838.1 | |||
| XM_017017350.1 | XP_016872839.1 | |||
| XM_017017351.1 | XP_016872840.1 | |||
| AY313782.1 | AAQ72549.1 | |||
| AY326450.1 | AAP92403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509434 | Chr.11:77115129 - 77231618 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map