Assay Details
Target Gene Details
Entrez Gene ID: | 5437 |
Gene Name: | RNA polymerase II subunit H |
Gene Aliases: |
RPABC3, RPB17, RPB8 |
Location: |
Chr.3:184361710-184368595 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2H | NM_001278698.1 | NP_001265627.1 | ||
| NM_001278699.1 | NP_001265628.1 | |||
| NM_001278700.1 | NP_001265629.1 | |||
| NM_001278714.1 | NP_001265643.1 | |||
| NM_001278715.1 | NP_001265644.1 | |||
| NM_006232.3 | NP_006223.2 | |||
| XM_006713666.2 | XP_006713729.1 | |||
| XM_006713667.2 | XP_006713730.1 | |||
| XM_006713668.3 | XP_006713731.1 | |||
| XM_006713670.2 | XP_006713733.1 | |||
| XM_017006636.1 | XP_016862125.1 | |||
| XM_017006637.1 | XP_016862126.1 | |||
| BC000739.2 | AAH00739.1 | |||
| BG529732.1 | ||||
| BI603667.1 | ||||
| BM541682.1 | ||||
| BQ065952.1 | ||||
| HY164762.1 | ||||
| U37689.1 | AAA91458.1 | |||
| Z49199.1 | CAA89060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422400 | Chr.3:184299495 - 184425713 on Build GRCh38 | Duplication |
|
| nsv829813 | Chr.3:184264997 - 184427996 on Build GRCh38 | Loss |
|
| nsv822363 | Chr.3:184364962 - 184366844 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map