Assay Details
Target Gene Details
Entrez Gene ID: | 2049 |
Gene Name: | EPH receptor B3 |
Gene Aliases: |
EK2, ETK2, HEK2, TYRO6 |
Location: |
Chr.3:184561799-184582408 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHB3 | NM_004443.3 | 1 | 99 | NP_004434.2 |
| BC052968.1 | 1 | 82 | AAH52968.1 | |
| CB993133.1 | 1 | 137 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598946 | Chr.3:184450977 - 184897567 on Build GRCh38 | Gain |
|
| nsv829814 | Chr.3:184404072 - 184622115 on Build GRCh38 | Loss |
|
| nsv508972 | Chr.3:184538711 - 184616364 on Build GRCh38 | Insertion |
|
| nsv461034 | Chr.3:184539010 - 184613658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map