Assay Details
Target Gene Details
Entrez Gene ID: | 10218 |
Gene Name: | angiopoietin like 7 |
Gene Aliases: |
AngX, CDT6, dJ647M16.1 |
Location: |
Chr.1:11189289-11195981 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL7 | NM_021146.3 | NP_066969.1 | ||
| AB593026.1 | ||||
| AB593027.1 | ||||
| AK313716.1 | ||||
| AY358301.1 | AAQ88668.1 | |||
| BC001881.1 | AAH01881.1 | |||
| BT009802.1 | AAP88804.1 | |||
| Y16132.1 | CAA76078.1 |
Target Gene Details
Entrez Gene ID: | 2475 |
Gene Name: | mechanistic target of rapamycin |
Gene Aliases: |
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS |
Location: |
Chr.1:11106531-11262551 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTOR | NM_004958.3 | NP_004949.1 | ||
| XM_005263438.2 | XP_005263495.1 | |||
| XM_011541166.2 | XP_011539468.1 | |||
| XM_017000900.1 | XP_016856389.1 | |||
| XM_017000901.1 | XP_016856390.1 | |||
| XM_017000902.1 | XP_016856391.1 | |||
| AB209995.1 | ||||
| AK302863.1 | ||||
| AK304273.1 | ||||
| BC117166.1 | ||||
| L34075.1 | AAA58486.1 | |||
| U88966.1 | AAC39933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map