Assay Details
Target Gene Details
Entrez Gene ID: | 51726 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B11 |
Gene Aliases: |
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537 |
Location: |
Chr.3:186570676-186585800 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB11 | NM_016306.5 | NP_057390.1 | ||
| AB028859.2 | BAA88307.1 | |||
| AF228505.1 | AAF61711.1 | |||
| AF277317.1 | AAK69110.1 | |||
| AJ250137.1 | CAB65118.1 | |||
| AK075300.1 | BAC11533.1 | |||
| AK075430.1 | BAC11617.1 | |||
| AK124289.1 | ||||
| AK301292.1 | ||||
| AY359043.1 | AAQ89402.1 | |||
| BC001144.1 | AAH01144.1 | |||
| BT007063.1 | AAP35712.1 | |||
| CR457096.1 | CAG33377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569155 | Chr.3:186578328 - 186588752 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map