Assay Details
Target Gene Details
Entrez Gene ID: | 23417 |
Gene Name: | malonyl-CoA decarboxylase |
Gene Aliases: |
MCD |
Location: |
Chr.16:83899125-83916182 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLYCD | NM_012213.2 | 1 | 437 | NP_036345.2 |
| AF090834.1 | 1 | 417 | AAD48994.1 | |
| AF097832.2 | 1 | 355 | AAD16177.2 | |
| AF153679.1 | 1 | 395 | AAD34631.1 | |
| BC000286.1 | 1 | 389 | AAH00286.1 | |
| BC052592.1 | 1 | 437 | AAH52592.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv573401 | Chr.16:83679394 - 83940461 on Build GRCh38 | Gain |
|
| nsv1908 | Chr.16:83876721 - 83909333 on Build GRCh38 | Insertion |
|
| nsv573398 | Chr.16:83516215 - 83940461 on Build GRCh38 | Gain |
|
| dgv3054n100 | Chr.16:83519838 - 83940461 on Build GRCh38 | Gain |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| nsv573406 | Chr.16:83817468 - 83908216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map