Assay Details
Target Gene Details
Entrez Gene ID: | 10930 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 2 |
Gene Aliases: |
ARCD1, ARP1 |
Location: |
Chr.6:41053201-41064891 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC2 | NM_006789.3 | 3 | 925 | NP_006780.1 |
| AF161698.1 | 3 | 833 | AAD45360.1 | |
| AI096570.1 | 1 | 336 | ||
| AK223461.1 | 3 | 925 | BAD97181.1 | |
| AK300689.1 | 4 | 699 | ||
| AK308197.1 | 3 | 756 | ||
| AK310006.1 | 5 | 737 | ||
| BC047767.2 | 3 | 862 | AAH47767.1 | |
| BC069688.1 | 3 | 822 | AAH69688.1 | |
| BC069764.1 | 3 | 825 | AAH69764.1 | |
| CB051273.1 | 1 | 360 |
More Information
Set Membership: |
|

Genomic Map