Assay Details
Target Gene Details
Entrez Gene ID: | 64432 |
Gene Name: | mitochondrial ribosomal protein S25 |
Gene Aliases: |
MRP-S25, RPMS25 |
Location: |
Chr.3:15042251-15065337 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS25 | NM_022497.4 | 4 | 4415 | NP_071942.1 |
| NR_135246.1 | ||||
| NR_135247.1 | ||||
| AI288348.1 | 1 | 188 | ||
| AK024433.1 | 4 | 4330 |
Target Gene Details
Entrez Gene ID: | 7182 |
Gene Name: | nuclear receptor subfamily 2 group C member 2 |
Gene Aliases: |
TAK1, TR4 |
Location: |
Chr.3:14947584-15049279 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2C2 | NM_001291694.1 | 14 | 7836 | NP_001278623.1 |
| NM_003298.4 | 15 | 7893 | NP_003289.2 | |
| XM_011534058.2 | 18 | 7928 | XP_011532360.1 | |
| XM_011534059.2 | 18 | 8834 | XP_011532361.1 | |
| XM_011534061.2 | 16 | 7782 | XP_011532363.1 | |
| XM_011534063.2 | 17 | 7869 | XP_011532365.1 | |
| XM_011534064.2 | 17 | 8031 | XP_011532366.1 | |
| XM_011534065.2 | 16 | 7741 | XP_011532367.1 | |
| XM_011534066.2 | 15 | 8544 | XP_011532368.1 | |
| XM_017007118.1 | 16 | 8010 | XP_016862607.1 | |
| XM_017007119.1 | 16 | 7670 | XP_016862608.1 | |
| XM_017007120.1 | 14 | 7678 | XP_016862609.1 | |
| AK091226.1 | 1 | 3141 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759130 | Chr.3:14869763 - 15121196 on Build GRCh38 | Gain |
|
| nsv834620 | Chr.3:14912162 - 15093316 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map