Assay Details
Target Gene Details
Entrez Gene ID: | 29760 |
Gene Name: | B-cell linker |
Gene Aliases: |
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca |
Location: |
Chr.10:96189171-96271576 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLNK | NM_001114094.1 | 16 | 1663 | NP_001107566.1 |
| NM_001258440.1 | 16 | 1516 | NP_001245369.1 | |
| NM_001258441.1 | 15 | 1447 | NP_001245370.1 | |
| NM_001258442.1 | 13 | 1201 | NP_001245371.1 | |
| NM_013314.3 | 17 | 1732 | NP_037446.1 | |
| NR_047680.1 | 16 | 1496 | ||
| NR_047681.1 | 15 | 1444 | ||
| NR_047682.1 | 15 | 1439 | ||
| NR_047683.1 | 14 | 1377 | ||
| XM_011539728.2 | 17 | 1609 | XP_011538030.1 | |
| XM_017016159.1 | 16 | 1541 | XP_016871648.1 | |
| AF068180.1 | 17 | 1707 | AAC39936.1 | |
| AF068181.1 | 16 | 1638 | AAC39937.1 | |
| AI307343.1 | 1 | 98 | ||
| AK027779.1 | 17 | 1667 | ||
| AK225546.1 | 16 | 1646 | ||
| BC018906.2 | 17 | 1646 | AAH18906.1 | |
| BF975580.1 | 5 | 663 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
| dgv29n17 | Chr.10:96187285 - 96197568 on Build GRCh38 | Loss |
|
| nsv831947 | Chr.10:96085409 - 96258168 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map