Assay Details
Target Gene Details
Entrez Gene ID: | 161424 |
Gene Name: | NOP9 nucleolar protein |
Gene Aliases: |
C14orf21 |
Location: |
Chr.14:24299854-24309126 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP9 | NM_001286367.1 | NP_001273296.1 | ||
| NM_174913.2 | NP_777573.1 | |||
| XM_005267385.1 | XP_005267442.1 | |||
| XM_011536526.1 | XP_011534828.1 | |||
| AB095941.1 | ||||
| AK123717.1 | ||||
| BC025332.1 | AAH25332.1 | |||
| BX248018.1 | CAD62343.1 | |||
| BX647311.1 | CAI46005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564058 | Chr.14:24302079 - 24317770 on Build GRCh38 | Loss |
|
| esv2669637 | Chr.14:24274158 - 24318169 on Build GRCh38 | Deletion |
|
| esv3633804 | Chr.14:24274134 - 24318147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map