Assay Details
Target Gene Details
Entrez Gene ID: | 925 |
Gene Name: | CD8a molecule |
Gene Aliases: |
CD8, Leu2, MAL, p32 |
Location: |
Chr.2:86784605-86808396 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD8A | NM_001145873.1 | 5 | 1241 | NP_001139345.1 |
| NM_001768.6 | 2 | 1099 | NP_001759.3 | |
| NM_171827.3 | 2 | 1099 | NP_741969.1 | |
| NR_027353.1 | 3 | 685 | ||
| AK097942.1 | 2 | 329 | ||
| AK124156.1 | 5 | 1241 | BAC85789.1 | |
| AK300089.1 | 3 | 651 | ||
| AY039664.1 | 2 | 389 | AAK72403.1 | |
| BC025715.1 | 2 | 215 | AAH25715.1 | |
| M12824.1 | 2 | 297 | AAA61133.1 | |
| M12828.1 | 2 | 275 | AAB04637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10063 | Chr.2:86713865 - 86857237 on Build GRCh38 | Gain |
|
| dgv714e214 | Chr.2:86745573 - 86951144 on Build GRCh38 | Gain |
|
| esv25494 | Chr.2:86745091 - 86946998 on Build GRCh38 | Gain |
|
| nsv582373 | Chr.2:86747195 - 86881088 on Build GRCh38 | Gain |
|
| nsv997982 | Chr.2:86745711 - 86878741 on Build GRCh38 | Gain |
|
| esv3591559 | Chr.2:86764344 - 86832450 on Build GRCh38 | Gain |
|
| dgv3886n100 | Chr.2:86721712 - 86945592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map