Assay Details
Target Gene Details
Entrez Gene ID: | 9854 |
Gene Name: | C2CD2 like |
Gene Aliases: |
DLNB23, TMEM24 |
Location: |
Chr.11:119107061-119118543 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD2L | NM_001290474.1 | 14 | 2697 | NP_001277403.1 |
| NM_014807.4 | 14 | 2700 | NP_055622.3 | |
| XM_006718948.3 | 13 | 2878 | XP_006719011.1 | |
| XM_011543076.2 | 14 | 3196 | XP_011541378.1 | |
| XM_017018625.1 | 13 | 2875 | XP_016874114.1 | |
| XM_017018626.1 | 13 | 2788 | XP_016874115.1 | |
| XM_017018627.1 | 13 | 2785 | XP_016874116.1 | |
| XM_017018628.1 | 14 | 2122 | XP_016874117.1 | |
| AB006623.2 | 14 | 2162 | BAA22954.2 | |
| AB094094.1 | 14 | 2668 | BAC76048.1 | |
| AK025223.1 | 14 | 2668 | ||
| BC003598.1 | 6 | 1182 | AAH03598.2 | |
| BC010071.2 | 8 | 1390 | AAH10071.2 | |
| BC021254.2 | 8 | 1390 | AAH21254.2 | |
| BC022219.1 | 14 | 2563 | AAH22219.1 | |
| BC033083.1 | 8 | 1390 | AAH33083.1 | |
| BC052246.1 | 6 | 1170 | AAH52246.1 | |
| BC075832.1 | 8 | 1390 | AAH75832.1 | |
| BX327704.2 | 1 | 71 | ||
| BX537572.1 | 7 | 5251 | ||
| CA309483.1 | 1 | 703 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv514 | Chr.11:119114552 - 119143217 on Build GRCh38 | Deletion |
|
| nsv468879 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map