Assay Details
Target Gene Details
Entrez Gene ID: | 9733 |
Gene Name: | squamous cell carcinoma antigen recognized by T-cells 3 |
Gene Aliases: |
DSAP1, P100, RP11-13G14, TIP110, p110, p110(nrb) |
Location: |
Chr.12:108522214-108561402 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SART3 | NM_014706.3 | 19 | 3814 | NP_055521.1 |
| XM_005269241.4 | 19 | 3881 | XP_005269298.1 | |
| XM_011539026.2 | 19 | 3349 | XP_011537328.1 | |
| AB020880.1 | 19 | 3598 | BAA78384.1 | |
| AB073647.1 | 4 | 1251 | ||
| AF387506.1 | 19 | 3582 | AAK69347.1 | |
| CR933631.1 | 12 | 8345 | ||
| D63879.1 | 19 | 3579 | BAA09929.1 |
More Information
Set Membership: |
|

Genomic Map