Assay Details
Target Gene Details
Entrez Gene ID: | 402055 |
Gene Name: | SRR1 domain containing |
Gene Aliases: |
HC/HCC, SRR1L |
Location: |
Chr.22:26483867-26494620 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRD | XM_011530178.2 | 7 | 1580 | XP_011528480.1 |
Target Gene Details
Entrez Gene ID: | 24144 |
Gene Name: | tuftelin interacting protein 11 |
Gene Aliases: |
NTR1, Spp382, TIP39, bK445C9.6, hNtr1 |
Location: |
Chr.22:26491927-26512496 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFIP11 | NM_001008697.1 | 16 | 2692 | NP_001008697.1 |
| NM_012143.2 | 15 | 2615 | NP_036275.1 | |
| XM_011530079.1 | 16 | 2675 | XP_011528381.1 | |
| XM_017028735.1 | 16 | 2833 | XP_016884224.1 | |
| AF070662.1 | 2 | 688 | AAD20968.1 | |
| AK025443.1 | 14 | 2739 | ||
| AK025739.1 | 11 | 2073 | ||
| AK298303.1 | 11 | 1675 | ||
| AK299964.1 | 10 | 1441 | ||
| AL050258.1 | 15 | 2628 | CAB43360.1 | |
| AL080147.1 | 15 | 2595 | CAB45740.1 | |
| BC011599.2 | 15 | 2592 | AAH11599.1 | |
| BC033080.1 | 15 | 2593 | AAH33080.1 | |
| BT007274.1 | 12 | 2365 | AAP35938.1 | |
| CT841511.2 | 13 | 2467 | CAJ86441.1 |
More Information
Set Membership: |
|

Genomic Map