Assay Details
Target Gene Details
Entrez Gene ID: | 26548 |
Gene Name: | integrin subunit beta 1 binding protein 2 |
Gene Aliases: |
CHORDC3, ITGB1BP, MELUSIN, MSTP015 |
Location: |
Chr.X:71301275-71305371 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB1BP2 | NM_001303277.1 | 1 | 84 | NP_001290206.1 |
| NM_012278.2 | 1 | 84 | NP_036410.1 | |
| XM_017029405.1 | 1 | 557 | XP_016884894.1 | |
| AF110225.1 | 1 | 38 | AAL36913.1 | |
| AF140690.1 | 1 | 54 | AAF01676.1 | |
| BC108901.2 | 1 | 45 | AAI08902.1 | |
| BP372847.1 | 1 | 82 | ||
| DA895605.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map