Assay Details
Target Gene Details
Entrez Gene ID: | 6879 |
Gene Name: | TATA-box binding protein associated factor 7 |
Gene Aliases: |
TAF2F, TAFII55 |
Location: |
Chr.5:141318490-141320784 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF7 | NM_005642.2 | 1 | 913 | NP_005633.2 |
| AK314837.1 | 1 | 896 | ||
| BC022798.1 | 1 | 891 | AAH22798.1 | |
| BC032737.1 | 1 | 871 | AAH32737.1 | |
| U18062.1 | 1 | 849 | AAB60347.1 | |
| X97999.1 | 1 | 896 | CAA66636.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map