Assay Details
Target Gene Details
Entrez Gene ID: | 9543 |
Gene Name: | immunoglobulin superfamily DCC subclass member 3 |
Gene Aliases: |
HsT18880, PUNC |
Location: |
Chr.15:65327127-65378040 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGDCC3 | NM_004884.3 | 12 | 2164 | NP_004875.2 |
| XM_011522241.1 | 12 | 2106 | XP_011520543.1 | |
| XM_011522243.1 | 11 | 1690 | XP_011520545.1 | |
| XM_011522244.1 | 11 | 1679 | XP_011520546.1 | |
| AF063936.1 | 4 | 396 | AAD13399.1 | |
| AK095529.1 | 11 | 1679 | ||
| BC042054.1 | 12 | 2112 | AAH42054.1 | |
| BC067107.1 | 12 | 2164 | AAH67107.1 | |
| BX389424.1 | 2 | 101 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569705 | Chr.15:65272257 - 65333881 on Build GRCh38 | Loss |
|
| nsv569706 | Chr.15:65293399 - 65337844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map