Assay Details
Target Gene Details
Entrez Gene ID: | 83692 |
Gene Name: | CD99 molecule like 2 |
Gene Aliases: |
CD99B, MIC2L1 |
Location: |
Chr.X:150766336-150898816 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD99L2 | NM_001184808.1 | 8 | 2821 | NP_001171737.1 |
| NM_001242614.1 | 12 | 3070 | NP_001229543.1 | |
| NM_031462.3 | 11 | 3040 | NP_113650.2 | |
| NM_134445.3 | 8 | 2824 | NP_604394.1 | |
| NM_134446.3 | 9 | 2893 | NP_604395.1 | |
| AK023776.1 | ||||
| AK024278.1 | 7 | 2547 | ||
| AK098810.1 | 1 | 1165 | ||
| AK125020.1 | 12 | 2881 | ||
| AK126859.1 | 2 | 3439 | ||
| AL136580.1 | 11 | 2929 | CAB66515.1 | |
| AY358837.1 | 11 | 2913 | AAQ89196.1 | |
| BC025729.1 | 12 | 4261 | AAH25729.1 | |
| BC030536.1 | 11 | 2900 | AAH30536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508813 | Chr.X:150735078 - 150811481 on Build GRCh38 | Insertion |
|
| nsv525661 | Chr.X:150731781 - 150792646 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map