Assay Details
Target Gene Details
Entrez Gene ID: | 51667 |
Gene Name: | negative regulator of ubiquitin like proteins 1 |
Gene Aliases: |
BS4, NUB1L, NYREN18 |
Location: |
Chr.7:151341699-151378461 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUB1 | NM_001243351.1 | 15 | 2162 | NP_001230280.1 |
| NM_016118.4 | 15 | 2120 | NP_057202.3 | |
| XM_005250011.2 | 15 | 2227 | XP_005250068.1 | |
| XM_017012304.1 | 15 | 2221 | XP_016867793.1 | |
| XM_017012305.1 | 15 | 2179 | XP_016867794.1 | |
| XM_017012306.1 | 15 | 2201 | XP_016867795.1 | |
| XM_017012307.1 | 14 | 2077 | XP_016867796.1 | |
| XM_017012308.1 | 14 | 2074 | XP_016867797.1 | |
| XM_017012309.1 | 14 | 2080 | XP_016867798.1 | |
| XM_017012310.1 | 16 | 2324 | XP_016867799.1 | |
| XM_017012311.1 | 16 | 2228 | XP_016867800.1 | |
| XM_017012312.1 | 16 | 2197 | XP_016867801.1 | |
| XM_017012313.1 | 16 | 2282 | XP_016867802.1 | |
| XM_017012314.1 | 16 | 2158 | XP_016867803.1 | |
| AF155099.1 | AAD42865.1 | |||
| AF300717.1 | 15 | 2109 | AAK21001.1 | |
| AF459743.1 | 15 | 2110 | AAO14547.1 | |
| AY129295.1 | 15 | 2068 | AAN01355.1 | |
| BC046354.2 | 14 | 2036 | AAH46354.1 |
More Information
Set Membership: |
|

Genomic Map