Assay Details
Target Gene Details
Entrez Gene ID: | 11309 |
Gene Name: | solute carrier organic anion transporter family member 2B1 |
Gene Aliases: |
OATP-B, OATP2B1, OATPB, SLC21A9 |
Location: |
Chr.11:75150987-75206400 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO2B1 | NM_001145211.2 | 5 | 626 | NP_001138683.1 |
| NM_001145212.2 | 2 | 499 | NP_001138684.1 | |
| NM_007256.4 | 5 | 931 | NP_009187.1 | |
| XM_017017157.1 | 5 | 549 | XP_016872646.1 | |
| AB020687.1 | 5 | 802 | ||
| AB026256.1 | 5 | 714 | BAA78638.1 | |
| AF205073.1 | 5 | 648 | AAG42205.1 | |
| AK290234.1 | 5 | 813 | ||
| AK294503.1 | 5 | 611 | ||
| AK300134.1 | 2 | 371 | ||
| AK302492.1 | 3 | 576 | ||
| AK304783.1 | ||||
| AL117465.1 | CAB55940.1 | |||
| BC041095.1 | 5 | 800 | AAH41095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054122 | Chr.11:75161747 - 75184196 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map