Assay Details
Target Gene Details
Entrez Gene ID: | 6240 |
Gene Name: | ribonucleotide reductase catalytic subunit M1 |
Gene Aliases: |
R1, RIR1, RR1 |
Location: |
Chr.11:4094685-4138925 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRM1 | NM_001033.4 | NP_001024.1 | ||
| NM_001318064.1 | NP_001304993.1 | |||
| AK222786.1 | BAD96506.1 | |||
| AK297988.1 | ||||
| AK299629.1 | ||||
| AK303394.1 | ||||
| AK307950.1 | ||||
| BC006498.2 | AAH06498.1 | |||
| BP229960.1 | ||||
| X59543.1 | CAA42118.1 | |||
| X59617.1 | CAA42180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578116 | Chr.11:4101889 - 4217176 on Build GRCh38 | Gain |
|
| nsv832053 | Chr.11:4075297 - 4309033 on Build GRCh38 | Gain |
|
| esv2759797 | Chr.11:4034715 - 4488958 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map