Assay Details
Target Gene Details
Entrez Gene ID: | 80344 |
Gene Name: | DDB1 and CUL4 associated factor 11 |
Gene Aliases: |
GL014, PRO2389, WDR23 |
Location: |
Chr.14:24114697-24125242 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF11 | NM_001163484.1 | 11 | 1248 | NP_001156956.1 |
| NM_025230.4 | 11 | 1698 | NP_079506.3 | |
| NM_181357.2 | 11 | 1050 | NP_852002.1 | |
| NR_028099.1 | 10 | 1610 | ||
| NR_028100.1 | 10 | 1225 | ||
| AB209073.1 | 10 | 1596 | BAD92310.1 | |
| AF130070.1 | 10 | 958 | AAG35498.1 | |
| AF267858.1 | 11 | 1361 | AAG44727.1 | |
| AK027223.1 | 4 | 227 | BAB15698.1 | |
| AK057636.1 | 11 | 1009 | ||
| AK074296.1 | 11 | 1663 | BAB85041.1 | |
| AK091857.1 | 2 | 1135 | ||
| AK094478.1 | ||||
| AK292797.1 | 11 | 1130 | ||
| AK296742.1 | 10 | 1182 | ||
| AK307849.1 | 8 | 830 | ||
| AL833831.1 | 4 | 215 | CAD38691.1 | |
| BC008858.2 | 6 | 476 | AAH08858.2 | |
| BC067132.1 | 11 | 1121 | AAH67132.1 | |
| BX161405.1 | 10 | 945 | CAD61885.1 | |
| BX247970.1 | 9 | 1124 | CAD62308.1 | |
| BX648224.1 | 11 | 1180 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map