Assay Details
Target Gene Details
Entrez Gene ID: | 27077 |
Gene Name: | B9 domain containing 1 |
Gene Aliases: |
B9, EPPB9, MKS9, MKSR1 |
Location: |
Chr.17:19335372-19378193 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B9D1 | NM_001243475.2 | 1 | 141 | NP_001230404.1 |
| XM_017024452.1 | 1 | 112 | XP_016879941.1 | |
| AK309320.1 | 1 | 130 | ||
| DB067258.1 | 1 | 141 |
Target Gene Details
Entrez Gene ID: | 5598 |
Gene Name: | mitogen-activated protein kinase 7 |
Gene Aliases: |
BMK1, ERK4, ERK5, PRKM7 |
Location: |
Chr.17:19377721-19383544 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK7 | NM_139032.2 | 1 | 333 | NP_620601.1 |
| NM_139033.2 | 1 | 333 | NP_620602.2 | |
| XM_006721558.3 | 1 | 333 | XP_006721621.1 | |
| AK295376.1 | ||||
| BC007404.2 | 1 | 301 | AAH07404.1 | |
| BC007992.2 | 1 | 301 | AAH07992.1 | |
| DB133227.1 | 1 | 333 | ||
| U29725.1 | 1 | 168 | AAA82931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv953845 | Chr.17:19377788 - 19387787 on Build GRCh38 | Deletion |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map